Vann, BAKER ANALYZED ULTRA LC/MS™, ULTRA for LC-MS, tested for UHPLC, J.T.Baker®

Leverandør: Avantor

Synonymer: Hydrogen oxide

Filtered through a 0.1 micron filter.

9823-02EA 2550 NOK
9823-02
Vann, BAKER ANALYZED ULTRA LC/MS™, ULTRA for LC-MS, tested for UHPLC, J.T.Baker®
Vann
Formel: H₂O
MW: 18.02 g/mol
Kokepunkt: 100
Smeltepunkt: 0
Tetthet: 1 g/cm³ (4 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011332
CAS-nummer: 7732-18-5
EINECS: 231-791-2
Merck Indeks: 13,10098

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance Passes Test
Gradient Elution Test (a.u.) - 254 nm ≤0.002
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.1 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤0.2 ppb
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) ≤25.0 ppb
LC/MS Suitability - Largest Response on ESI- Negative Mode (as 4-Nitrophenol) ≤25.0 ppb
Trace Impurities - Aluminum (Al) ≤10.0 ppb
Trace Impurities - Calcium (Ca) ≤20 ppb
Trace Impurities - Chromium (Cr) ≤5.0 ppb
Trace Impurities - Cobalt (Co) ≤5.0 ppb
Trace Impurities - Copper (Cu) ≤5.0 ppb
Trace Impurities - Iron (Fe) ≤5.0 ppb
Trace Impurities - Lead (Pb) ≤5.0 ppb
Trace Impurities - Lithium (Li) ≤5.0 ppb
Trace Impurities - Magnesium (Mg) ≤10.0 ppb
Trace Impurities - Manganese (Mn) ≤5.0 ppb
Trace Impurities - Nickel (Ni) ≤5.0 ppb
Trace Impurities - Potassium (K) ≤10.0 ppb
Trace Impurities - Sodium (Na) ≤20.0 ppb
Trace Impurities - Tin (Sn) ≤5.0 ppb
Trace Impurities - Zinc (Zn) ≤5.0 ppb
Filtered through a 0.1 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR