Vann, BAKER ANALYZED® for LC-MS, J.T.Baker®

Leverandør: Avantor

Synonymer: Hydrogen oxide

Management of Change (MOC) category = NI

9831-03EA 0 NOK
9831-03 9831-02
Vann, BAKER ANALYZED® for LC-MS, J.T.Baker®
Vann

For use in liquid chromatography and mass spectrometry.

Formel: H₂O
MW: 18.02 g/mol
Kokepunkt: 100
Smeltepunkt: 0
Tetthet: 1 g/cm³ (4 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011332
CAS-nummer: 7732-18-5
EINECS: 231-791-2
Merck Indeks: 13,10098

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance Passes Test
Gradient Elution Test (a.u.) - 254 nm ≤ 0.002
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤ 0.1 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤ 0.2 ppb
Color (APHA) ≤ 10
Residue after Evaporation ≤ 1.0 ppm
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) ≤ 50 ppb
Trace Impurities - Aluminum (Al) ≤ 50.0 ppb
Trace Impurities - * Calcium (Ca) ≤ 50.0 ppb
Trace Impurities - Iron (Fe) ≤ 50.0 ppb
Trace Impurities - Lithium (Li) ≤ 30.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 50.0 ppb
Trace Impurities - Nickel (Ni) ≤ 30.0 ppb
Trace Impurities - Potassium (K) ≤ 50 ppb
Trace Impurities - * Sodium (Na) ≤ 50.0 ppb
* May change over time due to extraction from glass container.
Filtered through a 0.2 micron filter.

SPESIFIKASJONER

Environmentally Preferable Safer Human and Environmental Health
Miljøsertifikat EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR