Tetrabutylammoniumhydrogensulfat concentrate for ion pair chromatography, Supelco®

Leverandør: Merck

Synonymer: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate

98479-6X1AMP-F
98479-6X1AMP-FEA 3260 NOK
98479-6X1AMP-F
Tetrabutylammoniumhydrogensulfat concentrate for ion pair chromatography, Supelco®
Tetrabutylammonium hydrogen sulphate

Dilute content from one ampule (~0.3M) to 1L with HPLC grade water.

Formel: (C₄H₉)₄NHSO₄
MDL nummer: MFCD00011637
CAS-nummer: 32503-27-8

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR