VWR®, Cetane Number standard, ISO 5165 / EN 61715
Leverandør: VWR Chemicals
99128.290EA 10300 NOK
99128.290
VWR®, Cetane Number standard, ISO 5165 / EN 61715
Standarder PetroleumIndustristandarder
Fuel testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Cetane Number ISO 5165 / EN 61715.
  • Certified in accordance with ASTM / EN or IP test method protocols
  • Certified by Round Robin method using independent test laboratories
  • Ensures good inter-laboratory correlation
  • Tamper-evident security pack