Gram's decolourising solution contains acetone and 2-propanol

Leverandør: VWR Chemicals

Synonymer: Gram's Decoloriser Solution

99502.5000EA 1550 NOK
99502.5000
Gram's decolourising solution contains acetone and 2-propanol
Gram's decolourising solution
MDL nummer: MFCD00240361
FN: 1993
ADR: 3 II

Bestill nå

Spesifikasjon Testresultater

Solution 50/50 (V/V) Clear liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR