VWR®, Density Standards, Certified Reference Materials, Referenced at 25 °C

Leverandør: VWR Chemicals

99656.600EA 1025 NOK
99656.600 99660.600
VWR®, Density Standards, Certified Reference Materials, Referenced at 25 °C
Standarder Tetthetsstandarder

VWR® ISO 17025 / ISO 17034 dual certified density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature.


  • ISO 17025 / ISO 17034 certified under UKAS accreditation
  • Density tested in strict accordance with ASTM D1480, a primary method for density determination
  • Density standards range between 0,6654 - 1,6300 g/ml
  • Ensures full compliance to ASTM and IP test method protocols
  • Shelf life between 12 / 24 months

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR