VWR®, Withdrawal System for Solvents with Manual Pressure Build-up for 10 and 30 L Stainless Steel Barrels
Leverandør: VWR Chemicals
99900.001EA 9275 NOK
99900.001
VWR®, Withdrawal System for Solvents with Manual Pressure Build-up for 10 and 30 L Stainless Steel Barrels
Pumper Fatpumper
Specially developed solvent withdrawal system with manual pressure build-up for VWR® returnable 10 and 30 L stainless steel barrels.
  • Two dip tubes
  • Hand pump ball
  • Perfluorelastomer o-rings
This set contains a specially developed solvent withdrawal system for manual pressure build-up which can be used with VWR® 10 and 30 L returnable stainless steel barrels. This set includes two dip tubes, one for 10 L and one for 30 L barrels as well as a hand pump ball with a brass coupling.
Leveringsinformasjon: Complete set contains withdrawal system for 10 and 3More Product Information