VWR®, Withdrawal System for 30 L Stainless Steel Barrels, Vertical
Leverandør: VWR Chemicals
99907.030EA 7050 NOK
99907.030
VWR®, Withdrawal System for 30 L Stainless Steel Barrels, Vertical
Pumper Fatpumper
Specially developed solvent withdrawal system for inert gas pressurizing.
  • Withdrawal system for 30 L barrels (stainless steel)
  • Perfluorelastomer o-rings
  • Includes 1 dip tube for 30 L barrels
Specially developed solvent withdrawal system for inert gas pressurizing for VWR® returnable 30 L stainless steel barrels.
Tilbehørs informasjon: Ideal for use with flexible hose with filling nozzle (99908.001).