Trikloreddiksyre (glasial) 20% in aqueous solution

Leverandør: PanReac AppliChem
Danger

Synonymer: TCA

A0590.1000
A0590.1000EA 1025 NOK
A0590.1000
Trikloreddiksyre (glasial) 20% in aqueous solution
Trikloreddiksyre (glasial)
Formel: Cl₃CCOOH
Kokepunkt: 196 °C (1013 hPa)
Smeltepunkt: 54…56 °C
Tetthet: ≤1,07 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004177
CAS-nummer: 76-03-9
EINECS: 200-927-2
FN: 2564
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR