Spermidine

Leverandør: PanReac AppliChem

Synonymer: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine

A0674.0005
A0674.0005EA 0 NOK
A0674.0005
Spermidine
Spermidine
Formel: C₇H₁₉N₃
Kokepunkt: 246 °C (1013 hPa)
Smeltepunkt: 20…25 °C
Tetthet: 0,925 g/cm³ (20 °C)
MDL nummer: MFCD00008229
CAS-nummer: 124-20-9
EINECS: 204-689-0
Merck Indeks: 12,08893

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR