Buffer, PBS (10X, Dulbecco's) powder pack
Leverandør: PanReac AppliChem
A0965.9010
A0965.9010EA 2725 NOK
A0965.9010
Buffer, PBS (10X, Dulbecco's) powder pack
Buffere Buffere, elektroforese Buffere, running
Powder mixture to prepare PBS buffer (10X) according to Dulbecco.
Composed of di-sodium hydrogen phosphate anhydrous (11500 mg/L), potassium chloride (2000 mg/L), potassium dihydrogen phosphate (2000 mg/L) and sodium chloride (80000 mg/L).