X-Gal

Leverandør: PanReac AppliChem

A1007.0001
A1007.0001EA 2825 NOK
A1007.0001
X-Gal
X-Gal
Formel: C₁₄H₁₅BrClNO₆
Smeltepunkt: 237…239 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00005666
CAS-nummer: 7240-90-6
EINECS: 230-640-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR