2,6-Dichlorophenolindophenol sodium salt hydrate

Leverandør: Thermo Fisher Scientific

A10107.06 A10107.14 A10107.22
A10107.06EA 915 NOK
A10107.06 A10107.14 A10107.22
2,6-Dichlorophenolindophenol sodium salt hydrate
2,6-Dichlorophenolindophenol sodium salt hydrate

2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase.


It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.

Formel: C₁₂H₆Cl₂NNaO₂·xH₂O
Smeltepunkt: >300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150014
CAS-nummer: 1266615-56-8
EINECS: 210-640-4
Merck Indeks: 14,3068

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR