Antracen ≥99%

Leverandør: Thermo Fisher Scientific
Warning

A10203.14 A10203.22 A10203.36
A10203.14EA 660 NOK
A10203.14 A10203.22 A10203.36
Antracen ≥99%
Antracen

Purity: 99% CAS No.: 120-12-7 Molecular Formula: C14H10 Formula Weight: 178.23 Flash Point: 121°(249°F) Melting Point: 214-218° Boiling Point: 340-342° Beilstein Reference: 1905429 Merck Reference: 14,682 UN No.: 3077 MDL No.: MFCD00001240

Formel: C₁₄H₁₀
Kokepunkt: 340…342 °C
Smeltepunkt: 214…218 °C
Tetthet: 1,250
Flash punkt: 121 °C (249 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001240
CAS-nummer: 120-12-7
EINECS: 204-371-1
ADR: 9,III
Merck Indeks: 14,682
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf973bfe5e591a235e84a414db5e3a617957ab78&vid=39774282d3251d5014028c1ba9d03866f87d7733&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR