Magnesiumnitrat heksahydrat ≥98%

Leverandør: Thermo Fisher Scientific
Warning

A10329.0B A10329.0I A10329.30
A10329.0BEA 885 NOK
A10329.0B A10329.0I A10329.30
Magnesiumnitrat heksahydrat ≥98%
Magnesiumnitrat heksahydrat

Purity: 98% CAS No.: 13446-18-9 Molecular Formula: Mg(NO3)2.6H2O Formula Weight: 256.41 (148.32anhy) Melting Point: 89° Boiling Point: 330° dec. Merck Reference: 14,5674 UN No.: 1474 MDL No.: MFCD00149779

Formel: Mg(NO₃)₂·6H₂O
Kokepunkt: 330 °C (dec.)
Smeltepunkt: 89 °C
Tetthet: 1,46
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149779
CAS-nummer: 13446-18-9
EINECS: 233-826-7
FN: 1474
ADR: 5.1,III
Merck Indeks: 14,5674

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR