Jern, pulver

Leverandør: Thermo Fisher Scientific
Warning

A10411.0I A10411.30 A10411.0B
A10411.0IEA 2850 NOK
A10411.0I A10411.30 A10411.0B
Jern, pulver
Jern
Formel: Fe
Kokepunkt: 3000 °C (1013 hPa)
Smeltepunkt: 1536 °C
Tetthet: 7,86 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010999
CAS-nummer: 7439-89-6
EINECS: 231-096-4
FN: 3089
ADR: 4.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR