Acetaldehyde oxime (mixture of syn and anti) 99%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Acetaldoxime

A10640.18 A10640.30
A10640.18EA 910 NOK
A10640.18 A10640.30
Acetaldehyde oxime (mixture of syn and anti) 99%
Acetaldehyde oxime (mixture of syn and anti)

Purity: 98% CAS No.: 107-29-9 Molecular Formula: CH3CH=NOH Formula Weight: 59.07 Flash Point: 36°(96°F) Refractive Index: 1.4260 Odour: Characteristic Boiling Point: 115-117° Beilstein Reference: 1209252 Merck Reference: 14,42 UN No.: 2332 MDL No.: MFCD00002124

Formel: C₂H₅NO
Kokepunkt: 115…117 °C
Tetthet: 0,969
Flash punkt: 36 °C (96 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002124
CAS-nummer: 107-29-9
EINECS: 203-479-6
FN: 2332
ADR: 3,III
Merck Indeks: 14,42

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR