Azomethine H sodium salt hydrate

Leverandør: Thermo Fisher Scientific
Warning

A10927.03 A10927.06 A10927.14
A10927.03EA 440 NOK
A10927.03 A10927.06 A10927.14
Azomethine H sodium salt hydrate
Azomethine H sodium salt hydrate

CAS No.: 206752-32-1 Molecular Formula: C17H12NNaO8S2.xH2O Formula Weight: 445.41(anhy) Melting Point: 300° Beilstein Reference: 4100175 MDL No.: MFCD00149588

Formel: C₁₇H₁₂NNaO₈S₂·xH₂O
Smeltepunkt: >300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149588
CAS-nummer: 206752-32-1
EINECS: 250-975-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR