Diphenyl sulphide ≥98%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Diphenylsulphane

A11114.14 A11114.22
A11114.14EA 605 NOK
A11114.14 A11114.22
Diphenyl sulphide ≥98%
Diphenyl sulphide

Purity: 98% CAS No.: 139-66-2 Molecular Formula: C12H10S Formula Weight: 186.28 Flash Point: 110°(230°F) Refractive Index: 1.6325 Odour: Unpleasant Melting Point: -40° Boiling Point: 295-296° Beilstein Reference: 1907932 Merck Reference: 14,7314 MDL No.: MFCD00003064

Formel: C₁₂H₁₀S
Kokepunkt: 295…296 °C
Smeltepunkt: –40 °C
Tetthet: 1,118
Flash punkt: >110 °C (230 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003064
CAS-nummer: 139-66-2
EINECS: 205-371-4
FN: 2810
ADR: 6.1,III
Merck Indeks: 14,7314

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR