3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: MCI-186, 1-Phenyl-3-methyl-5-pyrazolone, 3-Methyl-1-phenyl-1H-pyrazol-5(4H)-one, Edaravone

A11161.0E A11161.22 A11161.36
A11161.0EEA 3350 NOK
A11161.0E A11161.22 A11161.36
3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98%
3-Methyl-1-phenyl-2-pyrazolin-5-one

Purity: 98+% CAS No.: 89-25-8 Molecular Formula: C10H10N2O Formula Weight: 174.20 Odour: Odourless Melting Point: 127-131° Boiling Point: 191°/17mm Beilstein Reference: 609575 Merck Reference: 14,6713 MDL No.: MFCD00003138

Formel: C₁₀H₁₀N₂O
Kokepunkt: 191 °C (17 mmHg)
Smeltepunkt: 127…131 °C
Tetthet: 1,120
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003138
CAS-nummer: 89-25-8
EINECS: 201-891-0
Merck Indeks: 14,6713

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR