4-Nitrophenyl isocyanate ≥97%

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: 1-Isocyanato-4-nitrobenzene

A11194.03 A11194.06 A11194.14
A11194.03EA 415 NOK
A11194.03 A11194.06 A11194.14
4-Nitrophenyl isocyanate ≥97%
4-Nitrophenyl isocyanate

Purity: 97% CAS No.: 100-28-7 Molecular Formula: C7H4N2O3 Formula Weight: 164.12 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 56-59° Boiling Point: 137-138°/11mm Beilstein Reference: 389516 UN No.: 3335 MDL No.: MFCD00007306

Formel: C₇H₄N₂O₃
MW: 164.12 g/mol
Kokepunkt: 137…138 °C (11 mmHg)
Smeltepunkt: 56…59 °C
Flash punkt: >110 °C (230 °F)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007306
CAS-nummer: 100-28-7
EINECS: 202-836-3
FN: 3335
ADR: 9 III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR