Nikotinamid-adenin dinukleotid (oksidert form)

Leverandør: PanReac AppliChem

Synonymer: Nadide, NAD+, Nicotinamide adenine dinucleotide, oxidized

A1124.0005 A1124.0010 A1124.0025
A1124.0005EA 3275 NOK
A1124.0005 A1124.0010 A1124.0025
Nikotinamid-adenin dinukleotid (oksidert form)
Nikotinamid-adenin dinukleotid (oksidert form)
Formel: C₂₁H₂₇N₇O₁₄P₂
MW: 663.43 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036253
CAS-nummer: 53-84-9
EINECS: 200-184-4
Merck Indeks: 13,06370

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR