Etidiumbromid 10 mg/ml in aqueous solution

Leverandør: PanReac AppliChem
Danger

Synonymer: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide

A1152.0025
A1152.0025EA 725 NOK
A1152.0025
Etidiumbromid 10 mg/ml in aqueous solution
Etidiumbromid
Formel: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Kokepunkt: 72
MDL nummer: MFCD00011724
CAS-nummer: 1239-45-8
EINECS: 214-984-6
Merck Indeks: 13,04751

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR