Azure II

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: C.I. 52010/52015, 3,7 Bis(dimethylamino)phenothiazin-5-ium chloride and 3-(Dimethylamino)-7-(methylamino)-phenothiazin-5-ium chloride, Azur B/Methylene Blue 1:1, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride mixt. with 3-(dimethylamino)-7-(methylamino)phenothiazin-5-ium chloride(1:1), Methylene Azure II

A11777.09 A11777.18
A11777.09EA 520 NOK
A11777.09 A11777.18
Azure II
Azure II

CAS No.: 37247-10-2 MDL No.: MFCD00080716

Formel: C₁₆H₁₈N₃S·C₁₅H₁₆N₃S·2Cl
MW: 625.68 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00080716
CAS-nummer: 37247-10-2

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR