L(+)-Arabinose ≥99%

Leverandør: Thermo Fisher Scientific

A11921.18 A11921.30
A11921.18EA 1300 NOK
A11921.18 A11921.30
L(+)-Arabinose ≥99%
L(+)-Arabinose

Purity: 99% CAS No.: 87-72-9 Molecular Formula: C5H10O5 Formula Weight: 150.13 Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 155-160° Beilstein Reference: 1723085 Merck Reference: 14,761 Optical Rotation: +104° (c=10 in water, 24h) MDL No.: MFCD00067709

Formel: C₅H₁₀O₅
Kokepunkt: 331 °C (1013 hPa)
Smeltepunkt: 155…160 °C
Tetthet: 1,585 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00067709
CAS-nummer: 5328-37-0
EINECS: 201-767-6
Merck Indeks: 14,761
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad925ac21d4f12371621b1c44dbb6cda0a12da8&vid=6b9738c515009e5720388ea307490836c3b8afa8&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR