Antimon(III)klorid ≥99%

Leverandør: Thermo Fisher Scientific
Danger

A11944.0E A11944.22 A11944.36
A11944.0EEA 5525 NOK
A11944.0E A11944.22 A11944.36
Antimon(III)klorid ≥99%
Antimon(III)klorid

Purity: 99+% CAS No.: 10025-91-9 Molecular Formula: SbCl3 Formula Weight: 228.11 Odour: Pungent Melting Point: 73.4° Boiling Point: 223.5° Merck Reference: 14,707 UN No.: 1733 MDL No.: MFCD00011212

Formel: SbCl₃
Kokepunkt: 223,5 °C
Tetthet: 3,14
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011212
CAS-nummer: 10025-91-9
EINECS: 233-047-2
FN: 1733
ADR: 8,II
Merck Indeks: 14,707

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR