N,N-Dimetyl-p-toluidine ≥99%

Leverandør: Thermo Fisher Scientific
Danger

A11988.0E A11988.22 A11988.36
A11988.0EEA 7550 NOK
A11988.0E A11988.22 A11988.36
N,N-Dimetyl-p-toluidine ≥99%
N,N-Dimetyl-p-toluidine

Purity: 99% CAS No.: 99-97-8 Molecular Formula: C9H13N Formula Weight: 135.21 Flash Point: 83°(181°F) Refractive Index: 1.5460 Melting Point: -25° Boiling Point: 210-211° Beilstein Reference: 774409 UN No.: 1708 MDL No.: MFCD00008316

Formel: C₉H₁₃N
Kokepunkt: 210…211 °C
Smeltepunkt: –25 °C
Tetthet: 0,937
Flash punkt: 83 °C (181 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008316
CAS-nummer: 99-97-8
EINECS: 202-805-4
FN: 1708
ADR: 6.1,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR