1,1'-Dimethylferrocene ≥97%

Leverandør: Thermo Fisher Scientific

A12175.03 A12175.06 A12175.14
A12175.03EA 400 NOK
A12175.03 A12175.06 A12175.14
1,1'-Dimethylferrocene ≥97%
1,1'-Dimethylferrocene

Purity: 97% CAS No.: 1291-47-0 Molecular Formula: C12H14Fe Formula Weight: 214.09 Flash Point: 108°(226°F) Melting Point: 36-41° MDL No.: MFCD00001425

Formel: C₁₂H₁₄Fe
Smeltepunkt: 36…41 °C
Flash punkt: 108 °C (226 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001425
CAS-nummer: 1291-47-0
EINECS: 215-067-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR