Dimethyl succinate ≥98%

Leverandør: Thermo Fisher Scientific
Warning

A12565.0B A12565.22 A12565.30
A12565.0BEA 1675 NOK
A12565.0B A12565.22 A12565.30
Dimethyl succinate ≥98%
Dimethyl succinate

Purity: 98+% CAS No.: 106-65-0 Molecular Formula: C6H10O4 Formula Weight: 146.14 Flash Point: 85°(185°F) Refractive Index: 1.4190 Melting Point: 18-19° Boiling Point: 196° Beilstein Reference: 956776 Merck Reference: 14,8869 MDL No.: MFCD00008466

Formel: C₆H₁₀O₄
Kokepunkt: 196 °C
Smeltepunkt: 18…19 °C
Tetthet: 1,119
Flash punkt: 85 °C (185 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008466
CAS-nummer: 106-65-0
EINECS: 203-419-9
Merck Indeks: 14,8869

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR