4,6-Dichloro-o-toluidine ≥97%

Leverandør: Thermo Fisher Scientific
Danger

A12591.06 A12591.14
A12591.06EA 865 NOK
A12591.06 A12591.14
4,6-Dichloro-o-toluidine ≥97%
4,6-Dichloro-o-toluidine

Purity: 97% CAS No.: 30273-00-8 Molecular Formula: C7H7Cl2N Formula Weight: 176.05 Melting Point: 43-45° Boiling Point: 82-84°/0.1mm MDL No.: MFCD00044074

Formel: C₇H₇Cl₂N
Kokepunkt: 82…84 °C (0,1 mmHg)
Smeltepunkt: 43…45 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00044074
CAS-nummer: 30273-00-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR