4-Isopropoxybenzaldehyde ≥97%

Leverandør: Thermo Fisher Scientific
Warning

A12760.14 A12760.03 A12760.06
A12760.14EA 3575 NOK
A12760.14 A12760.03 A12760.06
4-Isopropoxybenzaldehyde ≥97%
4-Isopropoxybenzaldehyde

Purity: 97% CAS No.: 18962-05-5 Molecular Formula: C10H12O2 Formula Weight: 164.21 Refractive Index: 1.5470 Boiling Point: 108-110°/5mm MDL No.: MFCD00052357

Formel: C₁₀H₁₂O₂
Kokepunkt: 108…110 °C (5 mmHg)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00052357
CAS-nummer: 18962-05-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR