1H-1,2,4-Triazole-3-thiol ≥98%

Leverandør: Thermo Fisher Scientific
Danger

A13120.06 A13120.14 A13120.22
A13120.06EA 495 NOK
A13120.06 A13120.14 A13120.22
1H-1,2,4-Triazole-3-thiol ≥98%
1H-1,2,4-Triazole-3-thiol

Purity: 98% CAS No.: 3179-31-5 Molecular Formula: C2H3N3S Formula Weight: 101.13 Melting Point: 217-224° Beilstein Reference: 107731

Formel: C₂H₃N₃S
Smeltepunkt: 217…224 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005229
CAS-nummer: 3179-31-5
EINECS: 221-656-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR