1-Heksansulfonsyre natriumsalt ≥99%

Leverandør: Thermo Fisher Scientific

A13278.14 A13278.06 A13278.22
A13278.14EA 2775 NOK
A13278.14 A13278.06 A13278.22
1-Heksansulfonsyre natriumsalt ≥99%
1-Heksansulfonsyre natriumsalt

Purity: 99% CAS No.: 2832-45-3 Molecular Formula: CH3(CH2)5SO3Na Formula Weight: 188.22 Physical Form: Powder Melting Point: 300° Beilstein Reference: 3727014 MDL No.: MFCD00007542

Formel: C₆H₁₃O₃SNa
Smeltepunkt: >300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007542
CAS-nummer: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebc3cb7a7cefac9ad5a7678d82a90d244b508f9&vid=d608216fe1e698d6b206b03588e6d6fe5ee661e1&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR