Diethylamine hydrochloride 99%

Leverandør: Thermo Fisher Scientific
Warning

A13637.0B A13637.0I A13637.30
A13637.0BEA 1425 NOK
A13637.0B A13637.0I A13637.30
Diethylamine hydrochloride 99%
Diethylamine hydrochloride

Purity: 99% CAS No.: 660-68-4 Molecular Formula: (CH3CH2)2NH.HCl Formula Weight: 109.60 Melting Point: 227-230° Boiling Point: 320-330° Merck Reference: 14,3111 MDL No.: MFCD00012499

Formel: C₄H₁₂ClN
Kokepunkt: 320…330 °C
Smeltepunkt: 227…230 °C
Tetthet: 1,0475 g/cm³ (21 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012499
CAS-nummer: 660-68-4
EINECS: 211-541-9
Merck Indeks: 14,3111

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR