Antimony(III) fluoride ≥98%

Leverandør: Thermo Fisher Scientific
Danger

A14068.0B A14068.18 A14068.30
A14068.0BEA 4050 NOK
A14068.0B A14068.18 A14068.30
Antimony(III) fluoride ≥98%
Antimony(III) fluoride

Purity: 98% CAS No.: 7783-56-4 Molecular Formula: SbF3 Formula Weight: 178.75 Odour: Odourless Melting Point: 292° Boiling Point: 376° Merck Reference: 14,709 UN No.: 2923 MDL No.: MFCD00011218

Formel: F₃Sb
Kokepunkt: 376 °C
Smeltepunkt: 292 °C
Tetthet: 4,379
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011218
CAS-nummer: 7783-56-4
EINECS: 232-009-2
FN: 2923
ADR: 8,II
Merck Indeks: 14,709

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR