6-Aza-2-thiothymine 98%

Leverandør: Thermo Fisher Scientific

A14167.06 A14167.14 A14167.22
A14167.06EA 1050 NOK
A14167.06 A14167.14 A14167.22
6-Aza-2-thiothymine 98%
6-Aza-2-thiothymine

Purity: 98% CAS No.: 615-76-9 Molecular Formula: C4H5N3OS Formula Weight: 143.17 Melting Point: 218-221° Beilstein Reference: 4861 MDL No.: MFCD00006458

Formel: C₄H₅N₃OS
Smeltepunkt: 218…221 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006458
CAS-nummer: 615-76-9
EINECS: 210-445-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR