Erythrosin B disodium salt

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: C.I. 45430

A14180.14 A14180.22 A14180.36
A14180.14EA 675 NOK
A14180.14 A14180.22 A14180.36
Erythrosin B disodium salt
Erythrosin B disodium salt

CAS No.: 568-63-8 Molecular Formula: C20H6I4Na2O5 Formula Weight: 879.87 MDL No.: MFCD00144257


Acid Red 51

Formel: C₂₀H₆I₄Na₂O₅
MW: 879.86 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00144257
CAS-nummer: 16423-68-0
EINECS: 240-046-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR