L(+)-Leucin

Leverandør: PanReac AppliChem

Synonymer: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine

A1426.1000
A1426.1000EA 4425 NOK
A1426.1000
L(+)-Leucin
L(+)-Leucin
Formel: C₆H₁₃NO₂
MW: 131,17 g/mol
Smeltepunkt: 300 °C
Tetthet: 1,165 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002617
CAS-nummer: 61-90-5
EINECS: 200-522-0
Merck Indeks: 14,05451

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR