α,α,α-Trifluoro-p-toluic acid ≥98%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 4-(Trifluoromethyl)benzoic acid

A14494.06 A14494.14
A14494.06EA 845 NOK
A14494.06 A14494.14
α,α,α-Trifluoro-p-toluic acid ≥98%
α,α,α-Trifluoro-p-toluic acid

Purity: 98+% CAS No.: 455-24-3 Molecular Formula: C8H5F3O2 Formula Weight: 190.12 Melting Point: 220-222° Boiling Point: 247°/753mm MDL No.: MFCD00002562

Formel: C₈H₅F₃O₂
MW: 190.12 g/mol
Kokepunkt: 247 °C (753 mmHg)
Smeltepunkt: 220…222 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002562
CAS-nummer: 455-24-3
EINECS: 207-242-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR