cis-3-Chloroacrylic acid ≥98%

Leverandør: Thermo Fisher Scientific
Danger

A14798.03 A14798.06
A14798.03EA 705 NOK
A14798.03 A14798.06
cis-3-Chloroacrylic acid ≥98%
cis-3-Chloroacrylic acid

Purity: 98+% CAS No.: 1609-93-4 Molecular Formula: C3H3ClO2 Formula Weight: 106.51 Storage Temperature: 2°C to 8°C Melting Point: 60-62° Beilstein Reference: 1720278 UN No.: UN3261 MDL No.: MFCD00004368

Formel: C₃H₃ClO₂
Smeltepunkt: 60…62 °C
MDL nummer: MFCD00004368
CAS-nummer: 1609-93-4
EINECS: 216-545-4
ADR: 8,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR