Carbenicillin disodium salt

Leverandør: PanReac AppliChem
Danger

A1491.0005 A1491.0010
A1491.0005EA 2600 NOK
A1491.0005 A1491.0010
Carbenicillin disodium salt
Carbenicillin disodium salt
Formel: C₁₇H₁₆N₂O₆S·2Na
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00077683
CAS-nummer: 4800-94-6
EINECS: 225-360-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR