α,α,α-Trifluoro-o-toluidine ≥98%

Leverandør: Thermo Fisher Scientific
Danger

A15072.22 A15072.36
A15072.22EA 640 NOK
A15072.22 A15072.36
α,α,α-Trifluoro-o-toluidine ≥98%
α,α,α-Trifluoro-o-toluidine

Purity: 98% CAS No.: 88-17-5 Molecular Formula: C7H6F3N Formula Weight: 161.13 Flash Point: 65°(149°F) Refractive Index: 1.4800 Odour: Recognizable Melting Point: -35 to -33° Boiling Point: 67-68°/15mm Beilstein Reference: 879494 UN No.: 2942 MDL No.: MFCD00007718

Formel: C₇H₆F₃N
Kokepunkt: 67…68 °C (15 mmHg)
Smeltepunkt: –35…–33 °C
Tetthet: 1,295
Flash punkt: 65 °C (149 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007718
CAS-nummer: 88-17-5
EINECS: 201-806-7
FN: 2942
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR