EDTA dinatrium salt dihydrat ≥99%

Leverandør: Thermo Fisher Scientific

A15161.0B A15161.0I A15161.30
A15161.0BEA 1475 NOK
A15161.0B A15161.0I A15161.30
EDTA dinatrium salt dihydrat ≥99%
EDTA dinatrium salt dihydrat

Purity: 99+% CAS No.: 6381-92-6 Molecular Formula: C10H14N2Na2O8.2H2O Formula Weight: 372.24 (336.22anhy) Melting Point: 248° dec. Beilstein Reference: 3900609

Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Smeltepunkt: 248 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6
EINECS: 205-358-3
Merck Indeks: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR