4-Bromo-m-toluidine ≥97%

Leverandør: Thermo Fisher Scientific
Danger

A15417.09 A15417.18
A15417.09EA 375 NOK
A15417.09 A15417.18
4-Bromo-m-toluidine ≥97%
4-Bromo-m-toluidine

Purity: 97% CAS No.: 6933-10-4 Molecular Formula: C7H8BrN Formula Weight: 186.06 Odour: Characteristic Melting Point: 79-83° Boiling Point: 239-241° Beilstein Reference: 2689600 UN No.: 2811 MDL No.: MFCD00007828

Formel: C₇H₈BrN
Kokepunkt: 239…241 °C
Smeltepunkt: 79…83 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007828
CAS-nummer: 6933-10-4
EINECS: 230-056-3
FN: 2811
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR