Tetrabutylammoniumjodid ≥98%

Leverandør: Thermo Fisher Scientific
Warning

A15484.0B A15484.18 A15484.30
A15484.0BEA 4850 NOK
A15484.0B A15484.18 A15484.30
Tetrabutylammoniumjodid ≥98%
Tetrabutylammoniumjodid

Purity: 98% CAS No.: 311-28-4 Molecular Formula: [CH3(CH2)3]4NI Formula Weight: 369.38 Melting Point: 142-148° Beilstein Reference: 3916152

Formel: C₁₆H₃₆IN
Smeltepunkt: 142…148 °C
Tetthet: 1,0027 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011636
CAS-nummer: 311-28-4
EINECS: 206-220-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR