Nikkel(II)nitrat heksahydrat ≥98%

Leverandør: Thermo Fisher Scientific
Danger

A15540.0B A15540.0I A15540.22 A15540.36 A15540.A4
A15540.0BEA 1575 NOK
A15540.0B A15540.0I A15540.22 A15540.36 A15540.A4
Nikkel(II)nitrat heksahydrat ≥98%
Nikkel(II)nitrat heksahydrat

Purity: 98% CAS No.: 13478-00-7 Molecular Formula: N2NiO6.6H2O Formula Weight: 290.80 (182.70anhy) Odour: Odourless Melting Point: 56.7° Boiling Point: 137° Merck Reference: 14,6513 UN No.: 2725 MDL No.: MFCD00149805

Formel: Ni(NO₃)₂·6H₂O
Kokepunkt: 137 °C
Tetthet: 2,05
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149805
CAS-nummer: 13478-00-7
EINECS: 236-068-5
FN: 2725
ADR: 5.1,III
Merck Indeks: 14,6513

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR