Triethanolamine hydrochloride ≥99%

Leverandør: Thermo Fisher Scientific

A15678.0B A15678.0I A15678.30
A15678.0BEA 2550 NOK
A15678.0B A15678.0I A15678.30
Triethanolamine hydrochloride ≥99%
Triethanolamine hydrochloride

Purity: 99+% CAS No.: 637-39-8 Molecular Formula: C6H15NO3.HCl Formula Weight: 185.65 Melting Point: 177-179° Beilstein Reference: 3909940 Merck Reference: 14,9665 MDL No.: MFCD00012596

Formel: C₆H₁₆ClNO₃
Smeltepunkt: 177…179 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012596
CAS-nummer: 637-39-8
EINECS: 211-284-2
Merck Indeks: 14,9665

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR