cis-Aconitic anhydride 98%

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: Cis-aconitic anhydride

A16006.03 A16006.06 A16006.14
A16006.03EA 610 NOK
A16006.03 A16006.06 A16006.14
cis-Aconitic anhydride 98%
cis-Aconitic anhydride

Purity: 98% CAS No.: 6318-55-4 Molecular Formula: C6H4O5 Formula Weight: 156.10 Storage Temperature: 2°C to 8°C Melting Point: 74-78° Beilstein Reference: 127575 UN No.: UN3261 MDL No.: MFCD00005524

Formel: C₆H₄O₅
MW: 156.093 g/mol
Smeltepunkt: 74…78 °C
MDL nummer: MFCD00005524
CAS-nummer: 6318-55-4
EINECS: 228-663-3
ADR: 8 III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR