cis-Aconitic acid tech. 90%, technical

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: (Z)-Prop-1-ene-1,2,3-tricarboxylic acid, (1Z)-Prop-1-ene-1,2,3-tricarboxylic acid, (Z)-Aconitic acid

A16010.14 A16010.03 A16010.06
A16010.14EA 9400 NOK
A16010.14 A16010.03 A16010.06
cis-Aconitic acid tech. 90%, technical
cis-Aconitic acid

Purity: 90% CAS No.: 585-84-2 Molecular Formula: C6H6O6 Formula Weight: 174.11 Storage Temperature: 2°C to 8°C Odour: Odourless MDL No.: MFCD00063184

Formel: C₆H₆O₆
MW: 174.11 g/mol
Smeltepunkt: 115…120 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00063184
CAS-nummer: 585-84-2
EINECS: 209-561-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR