Glyoxal 40% (w/w) in aqueous solution

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Ethandial

A16144.0D A16144.0F A16144.AE A16144.AP
A16144.0DEA 5000 NOK
A16144.0D A16144.0F A16144.AE A16144.AP
Glyoxal 40% (w/w) in aqueous solution
Glyoxal

CAS No.: 107-22-2 Molecular Formula: C2H2O2 Formula Weight: 58.04 Refractive Index: 1.4090 Melting Point: -14° Boiling Point: 104° Merck Reference: 14,4509 MDL No.: MFCD00006957

Formel: C₂H₂O₂
MW: 58.04 g/mol
Kokepunkt: 104 °C
Smeltepunkt: –14 °C
Tetthet: 1.271
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006957
CAS-nummer: 107-22-2
EINECS: 203-474-9
Merck Indeks: 14,4509

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR